2,3,5,6-Tetrachloro-4-(trifluoromethyl)benzoyl chloride ≥97%

Supplier: Matrix Scientific

083073-1G 083073-10G
102838-854EA 226.15 USD
102838-854 102838-856
2,3,5,6-Tetrachloro-4-(trifluoromethyl)benzoyl chloride ≥97%
2,3,5,6-Tetrachloro-4-(trifluoromethyl)benzoyl chloride

Hazard: Irritant

Formula: C₈Cl₅F₃O
MDL Number: MFCD04974482
CAS Number: 117338-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR