4-(Difluoromethoxy)benzoyl chloride ≥97%

Supplier: Matrix Scientific

083072-1G 083072-10G
102838-850EA 178.46 USD
102838-850 102838-852
4-(Difluoromethoxy)benzoyl chloride ≥97%
4-(Difluoromethoxy)benzoyl chloride

Hazard: Irritant

Formula: C₈H₅ClF₂O₂
MDL Number: MFCD00833410
CAS Number: 57320-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR