Dibenzyl-2-aminosuccinate ≥97%

Supplier: Matrix Scientific

083067-1G 083067-10G
102838-838EA 292.31 USD
102838-838 102838-840
Dibenzyl-2-aminosuccinate ≥97%
Dibenzyl-2-aminosuccinate

Hazard: Irritant

Formula: C₁₈H₁₉NO₄
MDL Number: MFCD02114806
CAS Number: 4079-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR