S-(N,N-Dimethylthiocarbamoyl)thioglycolic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-((Dimethylcarbamothioyl)thio)acetic acid, S-(N,N-Dimethylthiocarbamoyl)mercaptoacetic Acid

083059-1G 083059-10G
102838-814EA 184.62 USD
102838-814 102838-816
S-(N,N-Dimethylthiocarbamoyl)thioglycolic acid ≥97%
S-(N,N-Dimethylthiocarbamoyl)thioglycolic acid

Hazard: Irritant

Formula: C₅H₉NO₂S₂
MW: 179.25 g/mol
MDL Number: MFCD00021765
CAS Number: 4007-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR