4-(Phenylhydrazono)pentanoic acid ≥97%

Supplier: Matrix Scientific

083057-1G 083057-10G
102838-806EA 101.54 USD
102838-806 102838-808
4-(Phenylhydrazono)pentanoic acid ≥97%
4-(Phenylhydrazono)pentanoic acid

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₂
MDL Number: MFCD00059636
CAS Number: 588-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR