6-(Acetylamino)hexanoic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 6-Acetamidocaproic Acid, 6-Acetamidohexanoic acid

083050-1G 083050-10G
102838-782EA 30.77 USD
102838-782 102838-784
6-(Acetylamino)hexanoic acid ≥97%
6-(Acetylamino)hexanoic acid

Hazard: Irritant

Formula: C₈H₁₅NO₃
MW: 173.21 g/mol
MDL Number: MFCD00004424
CAS Number: 57-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR