Methanesulfonylacetic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(Methylsulphonyl)acetic acid

083048-5G 083048-25G
102838-772EA 203.08 USD
102838-772 102838-774
Methanesulfonylacetic acid ≥97%
Methanesulfonylacetic acid

Hazard: Irritant

Formula: C₃H₆O₄S
MW: 138.14 g/mol
MDL Number: MFCD00051843
CAS Number: 2516-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR