2,2-Dimethyl-4-pentenoic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2,2-Dimethylpent-4-enoic acid

083041-1G 083041-5G 083041-25G
102838-746EA 107.69 USD
102838-746 102838-748 102838-750
2,2-Dimethyl-4-pentenoic acid ≥97%
2,2-Dimethyl-4-pentenoic acid

Hazard: Irritant

Formula: C₇H₁₂O₂
MDL Number: MFCD00012304
CAS Number: 16386-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR