4-(Dimethylamino)-4-oxobutanoic acid ≥97%

Supplier: Matrix Scientific

083038-1G 083038-10G
102838-736EA 84.62 USD
102838-736 102838-738
4-(Dimethylamino)-4-oxobutanoic acid ≥97%
4-(Dimethylamino)-4-oxobutanoic acid

Hazard: Irritant

Formula: C₆H₁₁NO₃
MDL Number: MFCD00043505
CAS Number: 2564-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR