9H-Fluoren-9-ylideneacetic acid ≥97%

Supplier: Matrix Scientific

083037-1G 083037-10G
102838-732EA 306.15 USD
102838-732 102838-734
9H-Fluoren-9-ylideneacetic acid ≥97%
9H-Fluoren-9-ylideneacetic acid

Hazard: Irritant

Formula: C₁₅H₁₀O₂
MDL Number: MFCD00040930
CAS Number: 4425-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR