Phenyl-3-chloropropionate ≥97%

Supplier: Matrix Scientific

Synonyms: Phenyl 3-chloropropanoate, 3-Chloropropionic Acid Phenyl Ester

083033-1G 083033-10G 083033-50G
102838-712EA 72.31 USD
102838-712 102838-714 102838-716
Phenyl-3-chloropropionate ≥97%
Phenyl-3-chloropropionate

Hazard: Irritant

Formula: C₉H₉ClO₂
MW: 184.62 g/mol
MDL Number: MFCD00045298
CAS Number: 24552-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR