3-Bromopyruvic acid hydrate ≥97%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
Bromopyruvic acid hydrate, 3-Bromo-2-oxopropionic acid hydrate
083017-1G
083017-10G
083017-50G
102838-676EA
101.54
USD
102838-676
102838-678
102838-680
3-Bromopyruvic acid hydrate ≥97%
3-Bromopyruvic acid hydrate
Purity: 97%
CAS: 206860-50-6
MDL Number: MFCD00149107
Molecular Formula: C3H5BrO4
Molecular Weight: 184,98
Hazard: Irritant
Formula:
C₃H₃BrO₃.xH₂O MW: 166.96 g/mol (anhy) |
MDL Number:
MFCD00149107 CAS Number: 206860-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...