(R)-(-)-2-Hydroxyisovaleric acid ≥97%
Supplier: Matrix Scientific
Synonyms:
D-α-Hydroxyisovaleric acid, 2-Hydroxyisovaleric-acid, (R)-2-Hydroxy-3-methylbutanoic acid, D-Valic acid, (2R)-2-Hydroxy-3-methylbutanoic acid
083009-1G
083009-10G
083009-50G
102838-638EA
170.77
USD
102838-638
102838-640
102838-642
(R)-(-)-2-Hydroxyisovaleric acid ≥97%
(R)-(-)-2-Hydroxyisovaleric acid
Hazard: Irritant
Formula:
C₅H₁₀O₃ MW: 118.13 g/mol Melting Pt: 69 °C |
MDL Number:
MFCD00066442 CAS Number: 17407-56-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...