2,3-Dioxoindoline-7-carboxylic acid ≥97%

Supplier: Matrix Scientific

082979-1G 082979-10G
102838-572EA 184.62 USD
102838-572 102838-574
2,3-Dioxoindoline-7-carboxylic acid ≥97%
2,3-Dioxoindoline-7-carboxylic acid

Hazard: Irritant

Formula: C₉H₅NO₄
MDL Number: MFCD00612492
CAS Number: 25128-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR