5-Benzotriazolecarboxylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 1H-Benzo[d][1,2,3]triazole-5-carboxylic acid, 5-Carboxybenzotriazole

082968-1G 082968-10G 082968-50G
102838-546EA 56.92 USD
102838-546 102838-548 102838-550
5-Benzotriazolecarboxylic acid ≥97%
5-Benzotriazolecarboxylic acid

Hazard: Irritant

Formula: C₇H₅N₃O₂
MW: 163.14 g/mol
MDL Number: MFCD00012318
CAS Number: 23814-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR