3-Oxoindane-1-carboxylic acid ≥97%

Supplier: Matrix Scientific

082963-1G 082963-10G 082963-50G
102838-532EA 101.54 USD
102838-532 102838-534 102838-536
3-Oxoindane-1-carboxylic acid ≥97%
3-Oxoindane-1-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₈O₃
MW: 176.17 g/mol
MDL Number: MFCD00092224
CAS Number: 29427-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR