3-Chromanecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Chroman-3-carboxylic acid

082953-1G 082953-10G
102838-510EA 213.85 USD
102838-510 102838-512
3-Chromanecarboxylic acid ≥95%
3-Chromanecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₀O₃
MDL Number: MFCD03783554
CAS Number: 115822-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR