6-Hydroxy-2-naphthoic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 6-hydroxynaphthalene-2-carboxylic acid

082941-1G 082941-10G
102838-482EA 72.31 USD
102838-482 102838-484
6-Hydroxy-2-naphthoic acid ≥97%
6-Hydroxy-2-naphthoic acid

Hazard: Irritant

Formula: C₁₁H₈O₃
MW: 188.18 g/mol
MDL Number: MFCD00060070
CAS Number: 16712-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR