3-Methyl-1H-indene-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

082936-1G 082936-10G
102838-468EA 213.85 USD
102838-468 102838-470
3-Methyl-1H-indene-2-carboxylic acid ≥97%
3-Methyl-1H-indene-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₀O₂
MW: 174.2 g/mol
MDL Number: MFCD00044428
CAS Number: 34225-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR