Theophylline-7-acetic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
2-(1,3-Dimethyl-2,6-dioxo-1,2,3,6-tetrahydro-7H-purin-7-yl)acetic acid
082930-100G
082930-500G
102838-454EA
192.31
USD
102838-454
102838-456
Theophylline-7-acetic acid ≥97%
Theophylline-7-acetic acid
Hazard: Irritant
Formula:
C₉H₁₀N₄O₄ |
MDL Number:
MFCD00022832 CAS Number: 652-37-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...