4,7-Dimethylpyrazolo[5,1-c]-[1,2,4]triazine-3-carboxylic acid ≥97%

Supplier: Matrix Scientific

082928-1G 082928-10G
102838-448EA 184.62 USD
102838-448 102838-450
4,7-Dimethylpyrazolo[5,1-c]-[1,2,4]triazine-3-carboxylic acid ≥97%
4,7-Dimethylpyrazolo[5,1-c]-[1,2,4]triazine-3-carboxylic acid

Hazard: Irritant

Formula: C₈H₈N₄O₂
MW: 192.18 g/mol
MDL Number: MFCD00067873
CAS Number: 175137-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR