8-(Aminocarbonyl)-1-naphthoic acid ≥95%

Supplier: Matrix Scientific

082923-1G 082923-10G
102838-436EA 101.54 USD
102838-436 102838-438
8-(Aminocarbonyl)-1-naphthoic acid ≥95%
8-(Aminocarbonyl)-1-naphthoic acid

Hazard: Irritant

Formula: C₁₂H₉NO₃
MDL Number: MFCD00052020
CAS Number: 5811-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR