1-Isopropyl-1H-1,2,3-benzotriazole-5-carboxylic acid ≥97%

Supplier: Matrix Scientific

082919-1G 082919-10G
102838-426EA 184.62 USD
102838-426 102838-428
1-Isopropyl-1H-1,2,3-benzotriazole-5-carboxylic acid ≥97%
1-Isopropyl-1H-1,2,3-benzotriazole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₁N₃O₂
MW: 205.22 g/mol
MDL Number: MFCD01570664
CAS Number: 306935-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR