2,7-Dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid ≥97%

Supplier: Matrix Scientific

082918-1G 082918-10G
102838-422EA 156.92 USD
102838-422 102838-424
2,7-Dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid ≥97%
2,7-Dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid

Hazard: Irritant

Formula: C₉H₉N₃O₂
Melting Pt: 245…247 °C
MDL Number: MFCD00067901
CAS Number: 175201-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR