1-(4-Chlorophenyl)cyclohexanecarboxylic acid ≥97%

Supplier: Matrix Scientific

082914-1G 082914-10G
102838-412EA 72.31 USD
102838-412 102838-414
1-(4-Chlorophenyl)cyclohexanecarboxylic acid ≥97%
1-(4-Chlorophenyl)cyclohexanecarboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₅ClO₂
MDL Number: MFCD00019350
CAS Number: 58880-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR