1-Cyclohexene-1-carboxylic acid ≥97%

Supplier: Matrix Scientific

082912-1G 082912-10G
102838-406EA 112.31 USD
102838-406 102838-408
1-Cyclohexene-1-carboxylic acid ≥97%
1-Cyclohexene-1-carboxylic acid

Hazard: Irritant

Formula: C₇H₁₀O₂
MDL Number: MFCD00001545
CAS Number: 636-82-8
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR