4-Methoxycyclohexanecarboxylic acid ≥97%

Supplier: Matrix Scientific

082892-1G 082892-10G
102838-364EA 241.54 USD
102838-364 102838-366
4-Methoxycyclohexanecarboxylic acid ≥97%
4-Methoxycyclohexanecarboxylic acid

Hazard: Irritant

Formula: C₈H₁₄O₃
MW: 158.2 g/mol
MDL Number: MFCD00074992
CAS Number: 95233-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR