[(4R)-2,2-Dimethyl-5-oxo-1,3-dioxolan-4-yl]acetic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (R)-2-(2,2-Dimethyl-5-oxo-1,3-dioxolan-4-yl)acetic acid

082885-1G 082885-10G
102838-346EA 101.54 USD
102838-346 102838-348
[(4R)-2,2-Dimethyl-5-oxo-1,3-dioxolan-4-yl]acetic acid ≥97%
[(4R)-2,2-Dimethyl-5-oxo-1,3-dioxolan-4-yl]acetic acid

Hazard: Irritant

Formula: C₇H₁₀O₅
MDL Number: MFCD00211272
CAS Number: 113278-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR