4-Hydroxycyclohexanecarboxylic acid (cis and trans mixture) ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxycyclohexanecarboxylic acid

082882-1G 082882-10G
102838-336EA 156.92 USD
102838-336 102838-338
4-Hydroxycyclohexanecarboxylic acid (cis and trans mixture) ≥97%
4-Hydroxycyclohexanecarboxylic acid (cis and trans mixture)

Hazard: Irritant

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
MDL Number: MFCD00077974
CAS Number: 17419-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR