1-(4-Methoxyphenyl)cyclopropanecarboxylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(4-Methoxyphenyl)-1-cyclopropanecarboxylic acid

082873-1G 082873-10G
102838-308EA 55.38 USD
102838-308 102838-310
1-(4-Methoxyphenyl)cyclopropanecarboxylic acid ≥97%
1-(4-Methoxyphenyl)cyclopropanecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₂O₃
MW: 192.2112 g/mol
MDL Number: MFCD00019223
CAS Number: 16728-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR