1-Methylcyclopropanecarboxylic acid ≥97%

Supplier: Matrix Scientific

082856-1G 082856-10G
102838-268EA 113.85 USD
102838-268 102838-270
1-Methylcyclopropanecarboxylic acid ≥97%
1-Methylcyclopropanecarboxylic acid

Hazard: Irritant

Formula: C₅H₈O₂
MDL Number: MFCD00001290
CAS Number: 6914-76-7
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR