5-Methyl-2-phenyl-2H-1,2,3-triazole-4-carboxylic acid ≥97%

Supplier: Matrix Scientific

082850-1G 082850-10G
102838-254EA 143.08 USD
102838-254 102838-256
5-Methyl-2-phenyl-2H-1,2,3-triazole-4-carboxylic acid ≥97%
5-Methyl-2-phenyl-2H-1,2,3-triazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₉N₃O₂
MDL Number: MFCD00051653
CAS Number: 22300-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR