(±)-2-Piperazinecarboxylic acid dihydrochloride ≥97%
Supplier: Matrix Scientific
Synonyms:
Piperazine-2-carboxylic acid dihydrochloride
082768-1G
082768-10G
102838-082EA
127.69
USD
102838-082
102838-084
(±)-2-Piperazinecarboxylic acid dihydrochloride ≥97%
(±)-2-Piperazinecarboxylic acid dihydrochloride
Hazard: Irritant
Formula:
C₅H₁₂Cl₂N₂O₂ MW: 203.07 g/mol |
MDL Number:
MFCD00044803 CAS Number: 3022-15-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...