3-[4-(tert-Butoxycarbonyl)piperazin-1-yl]propanoic acid ≥97%

Supplier: Matrix Scientific

082765-1G 082765-10G
102838-072EA 226.15 USD
102838-072 102838-074
3-[4-(tert-Butoxycarbonyl)piperazin-1-yl]propanoic acid ≥97%
3-[4-(tert-Butoxycarbonyl)piperazin-1-yl]propanoic acid

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₄
MDL Number: MFCD03410257
CAS Number: 242459-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR