4-[(Benzyloxy)carbonyl]-1-(tert-butoxycarbonyl)piperazine-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 4-((Benzyloxy)carbonyl)-1-(tert-butoxycarbonyl)piperazine-2-carboxylic acid, Piperazine-2-carboxylic acid, N1-BOC N4-CBZ protected

082762-1G 082762-10G
102838-064EA 113.85 USD
102838-064 102838-066
4-[(Benzyloxy)carbonyl]-1-(tert-butoxycarbonyl)piperazine-2-carboxylic acid ≥97%
4-[(Benzyloxy)carbonyl]-1-(tert-butoxycarbonyl)piperazine-2-carboxylic acid

Hazard: Irritant/Refrigerate

Formula: C₁₈H₂₄N₂O₆
Melting Pt: 150…152 °C
MDL Number: MFCD02683055
CAS Number: 149057-19-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR