1-Formyl-4-piperidinecarboxylic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
1-Formylpiperidine-4-carboxylic acid, 1-Formylisonipecotic Acid
082741-1G
082741-10G
102838-020EA
200
USD
102838-020
102838-022
1-Formyl-4-piperidinecarboxylic acid ≥97%
1-Formyl-4-piperidinecarboxylic acid
Hazard: Irritant
Formula:
C₇H₁₁NO₃ MW: 157.17 g/mol |
MDL Number:
MFCD03425206 CAS Number: 84163-42-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...