4-(3,4-Dihydro-2H-1,5-benzodioxepin-7-yl)-4-oxobutanoic acid ≥97%

Supplier: Matrix Scientific

082722-1G 082722-10G
102837-978EA 298.46 USD
102837-978 102837-980
4-(3,4-Dihydro-2H-1,5-benzodioxepin-7-yl)-4-oxobutanoic acid ≥97%
4-(3,4-Dihydro-2H-1,5-benzodioxepin-7-yl)-4-oxobutanoic acid

Hazard: Irritant

Formula: C₁₃H₁₄O₅
MDL Number: MFCD00218805
CAS Number: 175136-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR