(±)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: 1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid hydrochloride

082717-1G 082717-10G
102837-966EA 184.62 USD
102837-966 102837-968
(±)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid hydrochloride ≥97%
(±)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₂ClNO₂
MW: 213.6608 g/mol
MDL Number: MFCD00012743
CAS Number: 41994-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR