6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrochloride ≥97%

Supplier: Matrix Scientific

082714-1G 082714-10G
102837-958EA 255.38 USD
102837-958 102837-960
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrochloride ≥97%
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆ClNO₄
MDL Number: MFCD06661901
CAS Number: 30740-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR