Isoquinoline-3-carboxylic acid monohydrate ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Azanaphthalene-3-carboxylic acid monohydrate, 3-Carboxyisoquinoline monohydrate, Isoquinoline-3-carboxylic acid hydrate

082712-1G 082712-10G
102837-950EA 241.54 USD
102837-950 102837-952
Isoquinoline-3-carboxylic acid monohydrate ≥98%
Isoquinoline-3-carboxylic acid monohydrate

Hazard: Irritant

Formula: C₁₀H₉NO₃
MW: 191.183 g/mol
MDL Number: MFCD00075137
CAS Number: 207399-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR