3-Hydroxy-2-methylquinoline-4-carboxylic acid ≥97%

Supplier: Matrix Scientific

082708-1G 082708-10G
102837-938EA 72.31 USD
102837-938 102837-940
3-Hydroxy-2-methylquinoline-4-carboxylic acid ≥97%
3-Hydroxy-2-methylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₉NO₃
MW: 203.2 g/mol
MDL Number: MFCD00044850
CAS Number: 117-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR