4-Hydroxy-7-(trifluoromethyl)quinoline-3-carboxylic acid ≥97%

Supplier: Matrix Scientific

082707-1G 082707-10G
102837-934EA 101.54 USD
102837-934 102837-936
4-Hydroxy-7-(trifluoromethyl)quinoline-3-carboxylic acid ≥97%
4-Hydroxy-7-(trifluoromethyl)quinoline-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₆F₃NO₃
MW: 257.17 g/mol
MDL Number: MFCD00006771
CAS Number: 574-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR