5-(Benzyloxy)-1H-indole-2-carboxylic acid ≥85%

Supplier: Matrix Scientific

082689-1G 082689-10G
102837-890EA 224.62 USD
102837-890 102837-892
5-(Benzyloxy)-1H-indole-2-carboxylic acid ≥85%
5-(Benzyloxy)-1H-indole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₃NO₃
Melting Pt: 190 °C
MDL Number: MFCD00047165
CAS Number: 6640-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR