5-Methoxy-2-methyl-3-indoleacetic acid ≥97%

Supplier: Matrix Scientific

Synonyms: Indomethacin Related Compound A

082688-1G 082688-10G
102837-886EA 241.54 USD
102837-886 102837-888
5-Methoxy-2-methyl-3-indoleacetic acid ≥97%
5-Methoxy-2-methyl-3-indoleacetic acid

Hazard: Irritant

Formula: C₁₂H₁₃NO₃
MW: 219.24 g/mol
MDL Number: MFCD00005618
CAS Number: 2882-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR