3-(3-Methyl-1H-indol-1-yl)propanoic acid ≥97%

Supplier: Matrix Scientific

082685-1G 082685-10G
102837-878EA 241.54 USD
102837-878 102837-880
3-(3-Methyl-1H-indol-1-yl)propanoic acid ≥97%
3-(3-Methyl-1H-indol-1-yl)propanoic acid

Hazard: Irritant

Formula: C₁₂H₁₃NO₂
MDL Number: MFCD00052088
CAS Number: 57662-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR