6-Hydroxypyridazine-3-carboxylic acid monohydrate ≥97%

Supplier: Matrix Scientific

Synonyms: 6-Hydroxy-3-pyridazinecarboxylic Acid Monohydrate, 1,6-Dihydro-6-oxopyridazine-3-carboxylic acid monohydrate

082654-1G 082654-10G
102837-814EA 156.92 USD
102837-814 102837-816
6-Hydroxypyridazine-3-carboxylic acid monohydrate ≥97%
6-Hydroxypyridazine-3-carboxylic acid monohydrate

Hazard: Irritant

Formula: C₅H₆N₂O₄
MW: 158.11 g/mol
MDL Number: MFCD09064936
CAS Number: 306934-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR