[(4,6-Dimethylpyrimidin-2-yl)thio]acetic acid monohydrate ≥97%

Supplier: Matrix Scientific

082652-1G 082652-10G
102837-808EA 127.69 USD
102837-808 102837-810
[(4,6-Dimethylpyrimidin-2-yl)thio]acetic acid monohydrate ≥97%
[(4,6-Dimethylpyrimidin-2-yl)thio]acetic acid monohydrate

Hazard: Irritant

Formula: C₈H₁₂N₂O₃S
MW: 216.26 g/mol
MDL Number: MFCD00150584
CAS Number: 55749-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR