Pyrazine-2,3-dicarboxylic acid monoamide ≥97%

Supplier: Matrix Scientific

082648-1G 082648-10G
102837-796EA 101.54 USD
102837-796 102837-798
Pyrazine-2,3-dicarboxylic acid monoamide ≥97%
Pyrazine-2,3-dicarboxylic acid monoamide

Hazard: Irritant

Formula: C₆H₅N₃O₃
MDL Number: MFCD00191679
CAS Number: 67367-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR