2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid ≥97%

Supplier: Matrix Scientific

082604-1G 082604-10G
102837-704EA 113.85 USD
102837-704 102837-706
2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid ≥97%
2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid

Hazard: Irritant

Formula: C₁₂H₁₁NO₃
MDL Number: MFCD00100005
CAS Number: 107367-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR