Methyl-1H-pyrrole-1-carboxylate ≥97%
Supplier: Matrix Scientific
082547-1G
082547-10G
082547-50G
102837-580EA
29.23
USD
102837-580
102837-582
102837-584
Methyl-1H-pyrrole-1-carboxylate ≥97%
Methyl-1H-pyrrole-1-carboxylate
Hazard: Irritant
Formula:
C₆H₇NO₂ Boiling Pt: 180°C/770mm Flash Pt: 60°C |
MDL Number:
MFCD00126403 CAS Number: 4277-63-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...